Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ qwikSLOT Mobile Security Shelving Unit, Chrome

qwikSLOT Mobile Security Shelving Units keep valuable items safe at your fingertips.

$2610.00 - $3230.00

Specifications

Autoclavable Not Autoclavable
Color Silver
Material Steel
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
For Use With (Equipment) qwikSLOT Shelving
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
13-460-264
View Documents
Metro™
SEC33ECQPL
40.75 x 21.5 in.
Each for $2,610.00
Add to cart
 
13-460-261
View Documents
Metro™
SEC35ECQPL
52.75 x 21.5 in.
Each for $3,010.00
Add to cart
 
13-460-262
View Documents
Metro™
SEC53ECQPL
40.75 x 27.25 in.
Each for $2,660.00
Add to cart
 
13-460-263
View Documents
Metro™
SEC55ECQPL
52.75 x 27.25 in.
Each for $3,070.00
Add to cart
 
13-460-265
View Documents
Metro™
SEC56ECQPL
65 x 27.25 in.
Each for $3,230.00
Add to cart
 
Description

Description

  • Allows up to 20 different codes and has a 5+ year battery life. (2) AAA batteries included.
  • Heavy-gauge open wire construction keeps contents visible at all times, making it easy to check inventory.
  • Each door opens 270 degrees and can be secured along the sides of the unit.
  • Shipped knocked down to save on freight costs.
  • Assemble in minutes.
Specifications

Specifications

Not Autoclavable
Steel
qwikSLOT Shelving
Electronic PIN Lock
Yes
Silver
All Laboratory and Clean Room Non-Autoclavable Applications
68 in.
2
2 Swivel, 2 Brake, Plated Horn
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit