Alfa Aesar™ (R)-(+)-1,1'-Bi(2-naphthylamine), 97%

Manufacturer: Alfa Aesar™ H2578503

Quantity 1g

 View more versions of this product

Catalog No. AAH2578503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18741-85-0
    Molecular Formula C20H16N2
    Formula Weight 284.36
    MDL Number MFCD00145204
    Beilstein 5279621
    Synonym (R)-BINAM; (R)-(+)-2,2'-Diamino-1,1'-binaphthyl
    Chemical Name or Material (R)-(+)-1,1'-Bi(2-naphthylamine)

    Specifications

    Formula Weight 284.36
    MDL Number MFCD00145204
    CAS 18741-85-0
    Quantity 1g
    Optical Rotation +150° (c=1 in Pyridine)
    Molecular Formula C20H16N2
    Synonym (R)-BINAM; (R)-(+)-2,2'-Diamino-1,1'-binaphthyl
    Melting Point 242° to 243°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 5279621
    Health Hazard 1 Warning
    Chemical Name or Material (R)-(+)-1,1'-Bi(2-naphthylamine)
    Assay 97%

    Description

    Applications
    (R)-(+)-1,1 ft.-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit