Alfa Aesar™ (R)-(+)-1-(2-Bromophenyl)ethanol, 98%

Manufacturer: Alfa Aesar™ H56003MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH56003MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76116-20-6
    Molecular Formula C8H9BrO
    Formula Weight 201.06
    MDL Number MFCD00216591
    Synonym (R)-(+)-2-Bromo-alpha-methylbenzyl alcohol
    Chemical Name or Material (R)-(+)-1-(2-Bromophenyl)ethanol

    Specifications

    Formula Weight 201.06
    Molecular Formula C8H9BrO
    MDL Number MFCD00216591
    CAS 76116-20-6
    Synonym (R)-(+)-2-Bromo-alpha-methylbenzyl alcohol
    Melting Point 54° to 56°C
    Health Hazard 1 Warning
    Quantity 250mg
    Optical Rotation +54° (c=1 in Chloroform)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material (R)-(+)-1-(2-Bromophenyl)ethanol
    Assay 98%

    Description

    Applications
    It is the most important dyestuff intermediate.

    Notes
    Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit