Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-(2-Furyl)ethyl acetate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass ampule with screw-cap
113471-32-2
C8H10O3
154.165
MFCD01863300
WOYASPSVTWXGJJ-ZCFIWIBFSA-N
r-1-furan-2-yl ethyl acetate,1r-1-furan-2-yl ethyl acetate,r---1-2-furyl ethyl acetate,acetic acid r-1-2-furyl ethyl ester
5702607
[(1R)-1-(furan-2-yl)ethyl] acetate
CC(C1=CC=CO1)OC(=O)C
113471-32-2
C8H10O3
154.165
MFCD01863300
WOYASPSVTWXGJJ-ZCFIWIBFSA-N
r-1-furan-2-yl ethyl acetate,1r-1-furan-2-yl ethyl acetate,r---1-2-furyl ethyl acetate,acetic acid r-1-2-furyl ethyl ester
5702607
[(1R)-1-(furan-2-yl)ethyl] acetate
CC(C1=CC=CO1)OC(=O)C
Density 1.0500g/mL
Specific Rotation Condition − 3.40 (20.00°C )
Color Colorless
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass ampule with screw-cap
Refractive Index 1.4575 to 1.4595 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 154.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material (R)-(-)-1-(2-Furyl)ethyl acetate, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE