Alfa Aesar™ (R)-1-(3,4-Dimethoxyphenyl)ethylamine, ChiPros™, 98%, ee 98+%

Manufacturer: Alfa Aesar™ H2748406

Quantity 5g

 View more versions of this product

Catalog No. AAH2748406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100570-24-9
    Molecular Formula C10H15NO2
    Formula Weight 181.24
    MDL Number MFCD06761897
    UN Number UN2735
    Synonym (R)-3,4-Dimethoxy-alpha-methylbenzylamine
    Chemical Name or Material (R)-1-(3,4-Dimethoxyphenyl)ethylamine

    Specifications

    Formula Weight 181.24
    Molecular Formula C10H15NO2
    MDL Number MFCD06761897
    UN Number UN2735
    Synonym (R)-3,4-Dimethoxy-alpha-methylbenzylamine
    Boiling Point 120°C (1mmHg)
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    CAS 100570-24-9
    Health Hazard 1 Danger
    Chemical Name or Material (R)-1-(3,4-Dimethoxyphenyl)ethylamine
    Assay 98%, ee ≥98%

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit