Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(-)-1,3-Butanediol, 98%

Click to view available options
Quantity:
1g
5g
6290-03-5
C4H10O2
90.12
MFCD00064277
PUPZLCDOIYMWBV-UHFFFAOYNA-N
r---1,3-butanediol,r-butane-1,3-diol,3r-butane-1,3-diol,r-1,3-butanediol,1,3-butanediol, 3r,r---1,3-butylene glycol,r---butane-1,3-diol,1,3-butanediol, r
637497
CHEBI:52687
(3R)-butane-1,3-diol
CC(CCO)O
6290-03-5
C4H10O2
90.12
MFCD00064277
PUPZLCDOIYMWBV-UHFFFAOYNA-N
r---1,3-butanediol,r-butane-1,3-diol,3r-butane-1,3-diol,r-1,3-butanediol,1,3-butanediol, 3r,r---1,3-butylene glycol,r---butane-1,3-diol,1,3-butanediol, r
637497
CHEBI:52687
(3R)-butane-1,3-diol
CC(CCO)O
Applications
(R)-(-)-1,3-Butanediol is used as a solvent for food flavoring agents and is a co-monomer used in certain polyurethane and polyester resins.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Flash Point 121°C (249°F)
Refractive Index 1.44
Sensitivity Hygroscopic
Density 1.005
Quantity 5g
Optical Rotation −30° (Neat)
Boiling Point 203°C to 204°C
Odor Weak
Beilstein 1718944
Solubility Information Fully miscible in water.
Formula Weight 90.12
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,3-Butanediol
Show More Show Less

EINECSNumber : 228-532-0

TSCA : No

Recommended Storage : Ambient temperatures