Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Alfa Aesar™ (R)-(+)-1-(4-Pyridyl)ethanol, 99%

Manufacturer:  Alfa Aesar™ H51033-03

Quantity 1g

 View more versions of this product

Catalog No. AAH5103303

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

27854-88-2
123.15
4306037
(R)-(+)-1-(4-Pyridyl)ethanol
C7H9NO
MFCD00077865
(R)-(+)-alpha-Methyl-4-pyridinemethanol

Specifications

27854-88-2
123.15
4306037
1g
99%
C7H9NO
MFCD00077865
(R)-(+)-alpha-Methyl-4-pyridinemethanol
(R)-(+)-1-(4-Pyridyl)ethanol

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit