Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(r)-(-)-1-(α-aminobenzyl)-2-naphthol, 99%, Acros Organics

Click to view available options
Quantity:
1g
219897-35-5
C17H15NO
249.313
MFCD04972352
PZMIGEOOGFFCNT-QGZVFWFLSA-N
r---1-alpha-aminobenzyl-2-naphthol,unii-c44563v2gb,1-r-amino phenyl methyl naphthalen-2-ol,1-r-amino phenyl methyl naphthalen-2-ol,r-betti base,betti base, r,betti base,-,betti base r-form mi,alphachiron 228785a443,--1-alpha-aminobenzyl-2-naphthol
6935961
1-[(R)-amino(phenyl)methyl]naphthalen-2-ol
C1=CC=C(C=C1)C(C2=C(C=CC3=CC=CC=C32)O)N
219897-35-5
C17H15NO
249.313
MFCD04972352
PZMIGEOOGFFCNT-QGZVFWFLSA-N
r---1-alpha-aminobenzyl-2-naphthol,unii-c44563v2gb,1-r-amino phenyl methyl naphthalen-2-ol,1-r-amino phenyl methyl naphthalen-2-ol,r-betti base,betti base, r,betti base,-,betti base r-form mi,alphachiron 228785a443,--1-alpha-aminobenzyl-2-naphthol
6935961
1-[(R)-amino(phenyl)methyl]naphthalen-2-ol
C1=CC=C(C=C1)C(C2=C(C=CC3=CC=CC=C32)O)N
Color White
Infrared Spectrum Authentic
Specific Rotation Condition −55° (20°C c=2,benzene)
Quantity 1g
Specific Rotation -52° to -58° (20°C, 589nm) (c=2, benzene)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Merck Index 15, 1190
Formula Weight 249.31
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material (R)-(-)-1-(α-Aminobenzyl)-2-naphthol
Show More Show Less