Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Benzyl-3-pyrrolidinol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
101930-07-8
C11H15NO
177.25
YQMXOIAIYXXXEE-LLVKDONJSA-N
r-+-1-benzyl-3-pyrrolidinol,r-3-hydroxy-1-benzylpyrrolidine,3r-1-benzylpyrrolidin-3-ol,r-1-benzyl-3-pyrrolidinol,r-1-benzylpyrrolidin-3-ol,r-1-n-benzyl-3-hydroxypyrrolidine,r-1-benzyl-3-hydroxypyrrolidine,r-n-benzyl-3-hydroxypyrrolidine,r-1-benzyl-pyrrolidin-3-ol,r-bhp
643472
(3R)-1-benzylpyrrolidin-3-ol
C1CN(CC1O)CC2=CC=CC=C2
101930-07-8
C11H15NO
177.25
YQMXOIAIYXXXEE-LLVKDONJSA-N
r-+-1-benzyl-3-pyrrolidinol,r-3-hydroxy-1-benzylpyrrolidine,3r-1-benzylpyrrolidin-3-ol,r-1-benzyl-3-pyrrolidinol,r-1-benzylpyrrolidin-3-ol,r-1-n-benzyl-3-hydroxypyrrolidine,r-1-benzyl-3-hydroxypyrrolidine,r-n-benzyl-3-hydroxypyrrolidine,r-1-benzyl-pyrrolidin-3-ol,r-bhp
643472
(3R)-1-benzylpyrrolidin-3-ol
C1CN(CC1O)CC2=CC=CC=C2
Specific Rotation + 4
Boiling Point 88°C to 90°C (1.0 Torr)
Quantity 1g
Assay Percent Range 97.5% min. (GC)
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material (R)-(+)-1-Benzyl-3-pyrrolidinol

GHS Signal Word: Warning