Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-1-Benzyloxycarbonyl-5-pyrrolidinone-2-carboxylic acid, 95%

Click to view available options
Quantity:
5g
25g
78339-57-8
C13H13NO5
263.249
MFCD00077036
VHSFUGXCSGOKJX-SNVBAGLBSA-N
z-d-pyr-oh,cbz-d-pyr-oh,r-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid,2r-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid,ambotzzaa1212,benzyloxycarbonyl-d-pyroglutamic acid,r-n-alpha-benzyloxycarbonyl-pyroglutamic acid,2r-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid,1,2-pyrrolidinedicarboxylicacid, 5-oxo-, 1-phenylmethyl ester, 2r
688429
(2R)-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1CC(=O)N(C1C(=O)O)C(=O)OCC2=CC=CC=C2
78339-57-8
C13H13NO5
263.249
MFCD00077036
VHSFUGXCSGOKJX-SNVBAGLBSA-N
z-d-pyr-oh,cbz-d-pyr-oh,r-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid,2r-1-benzyloxy carbonyl-5-oxopyrrolidine-2-carboxylic acid,ambotzzaa1212,benzyloxycarbonyl-d-pyroglutamic acid,r-n-alpha-benzyloxycarbonyl-pyroglutamic acid,2r-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid,1,2-pyrrolidinedicarboxylicacid, 5-oxo-, 1-phenylmethyl ester, 2r
688429
(2R)-5-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1CC(=O)N(C1C(=O)O)C(=O)OCC2=CC=CC=C2
Quantity 25g
Color White
Melting Point 138°C to 144°C
Beilstein 4698210
Formula Weight 263.25
Physical Form Powder
Percent Purity 95%
Chemical Name or Material (R)-1-Benzyloxycarbonyl-5-pyrrolidinone-2-carboxylic acid

storageNote1 : -30° to -10°C