Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine, 97%

Click to view available options
Quantity:
250mg
1g
199174-29-3
C10H20N2O2
200.282
MFCD03844734
OGCCBDIYOAFOGK-MRVPVSSYSA-N
r-1-boc-3-aminomethyl pyrrolidine,r-n-boc-3-aminomethyl pyrrolidine,r-tert-butyl 3-aminomethyl pyrrolidine-1-carboxylate,r-3-aminomethyl-1-n-boc-pyrrolidine,tert-butyl 3r-3-aminomethyl pyrrolidine-1-carboxylate,r-1-n-boc-3-aminomethyl pyrrolidine,r-1-boc-3-aminomethylpyrrolidine,3r-3-aminomethyl-1-boc-pyrrolidine,r-3-aminomethyl-1-n-boc-pyrrolidine,r---1-boc-3-aminomethyl pyrrolidine
1515152
tert-butyl (3R)-3-(aminomethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)CN
199174-29-3
C10H20N2O2
200.282
MFCD03844734
OGCCBDIYOAFOGK-MRVPVSSYSA-N
r-1-boc-3-aminomethyl pyrrolidine,r-n-boc-3-aminomethyl pyrrolidine,r-tert-butyl 3-aminomethyl pyrrolidine-1-carboxylate,r-3-aminomethyl-1-n-boc-pyrrolidine,tert-butyl 3r-3-aminomethyl pyrrolidine-1-carboxylate,r-1-n-boc-3-aminomethyl pyrrolidine,r-1-boc-3-aminomethylpyrrolidine,3r-3-aminomethyl-1-boc-pyrrolidine,r-3-aminomethyl-1-n-boc-pyrrolidine,r---1-boc-3-aminomethyl pyrrolidine
1515152
tert-butyl (3R)-3-(aminomethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)CN
Quantity 1g
Color Colorless to Yellow
Refractive Index 1.4780
Sensitivity Air Sensitive
Beilstein 10537871
Formula Weight 200.28
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine

storageNote1 : 2° to 8°C