Alfa Aesar™ (R)-(+)-1-Boc-3-aminopyrrolidine, 97%

Manufacturer: Alfa Aesar™ H2818806

Quantity 5g

 View more versions of this product

Catalog No. AAH2818806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 147081-49-0
    Molecular Formula C9H18N2O2
    Formula Weight 186.25
    MDL Number MFCD03419272
    UN Number UN2735
    Synonym (R)-(+)-1-tert-Butoxycarbonyl-3-aminopyrrolidine; tert-Butyl (R)-(+)-3-aminopyrrolidine-1-carboxylate
    Chemical Name or Material (R)-(+)-1-Boc-3-aminopyrrolidine

    Specifications

    Formula Weight 186.25
    Boiling Point 243° to 244°C
    Flash Point 91°C (196°F)
    Optical Rotation +3.5° (c=1 in Chloroform)
    Molecular Formula C9H18N2O2
    MDL Number MFCD03419272
    CAS 147081-49-0
    UN Number UN2735
    Synonym (R)-(+)-1-tert-Butoxycarbonyl-3-aminopyrrolidine; tert-Butyl (R)-(+)-3-aminopyrrolidine-1-carboxylate
    Health Hazard 1 Danger
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (R)-(+)-1-Boc-3-aminopyrrolidine
    Assay 97%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit