Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(-)-1-Boc-3-fluoropyrrolidine, 96%

Click to view available options
Quantity:
250mg
1g
876617-25-3
C9H16FNO2
189.23
MFCD04114065
SUECTKVSIDXQQE-SSDOTTSWSA-N
r-1-boc-3-fluoropyrrolidine,r-tert-butyl 3-fluoropyrrolidine-1-carboxylate,tert-butyl 3r-3-fluoropyrrolidine-1-carboxylate,1-pyrrolidinecarboxylic acid, 3-fluoro-, 1,1-dimethylethyl ester, 3r,tert-butyl r-3-fluoropyrrolidine-1-carboxylate,r-1-boc-3-fluoropyrrolidine,r---1-boc-3-fluoropyrrolidine
40430177
tert-butyl (3R)-3-fluoropyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)F
876617-25-3
C9H16FNO2
189.23
MFCD04114065
SUECTKVSIDXQQE-SSDOTTSWSA-N
r-1-boc-3-fluoropyrrolidine,r-tert-butyl 3-fluoropyrrolidine-1-carboxylate,tert-butyl 3r-3-fluoropyrrolidine-1-carboxylate,1-pyrrolidinecarboxylic acid, 3-fluoro-, 1,1-dimethylethyl ester, 3r,tert-butyl r-3-fluoropyrrolidine-1-carboxylate,r-1-boc-3-fluoropyrrolidine,r---1-boc-3-fluoropyrrolidine
40430177
tert-butyl (3R)-3-fluoropyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)F
Quantity 1g
Sensitivity Moisture Sensitive
Boiling Point 85°C to 95°C (2mmHg)
Color Colorless to Yellow
Refractive Index 1.4420
UN Number UN2810
Formula Weight 189.23
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material (R)-(-)-1-Boc-3-fluoropyrrolidine

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.