Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%

Manufacturer:  Alfa Aesar™ H2690203

Quantity 1g

 View more versions of this product

Catalog No. AAH2690203


  Check Availability
Add to cart

Specifications

Description

Applications
(R)-1-Cyclopropylethylamine is used for resolution of organic compounds in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.
Description & Specifications

Chemical Identifiers

6240-96-6
85.15
UN2733
(R)-1-Cyclopropylethylamine
C5H11N
MFCD08064288
(R)-alpha-Methylcyclopropanemethylamine

Specifications

6240-96-6
85.15
UN2733
−68°C
95°C
3°C (37°F)
(R)-1-Cyclopropylethylamine
C5H11N
MFCD08064288
(R)-alpha-Methylcyclopropanemethylamine
13
1g
Amine-like
98%, ee ≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit