Alfa Aesar™ (R)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%

Manufacturer: Alfa Aesar™ H2690206

Quantity 5g

 View more versions of this product

Catalog No. AAH2690206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6240-96-6
    Molecular Formula C5H11N
    Formula Weight 85.15
    MDL Number MFCD08064288
    UN Number UN2733
    Synonym (R)-alpha-Methylcyclopropanemethylamine
    Chemical Name or Material (R)-1-Cyclopropylethylamine

    Specifications

    Formula Weight 85.15
    Molecular Formula C5H11N
    Melting Point −68°C
    pH 13
    Quantity 5g
    Flash Point 3°C (37°F)
    Odor Amine-like
    MDL Number MFCD08064288
    CAS 6240-96-6
    UN Number UN2733
    Synonym (R)-alpha-Methylcyclopropanemethylamine
    Health Hazard 1 Danger
    Boiling Point 95°C
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Store in a well-ventilated place.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Chemical Name or Material (R)-1-Cyclopropylethylamine
    Assay 98%, ee ≥98%

    Description

    Applications
    (R)-1-Cyclopropylethylamine is used for resolution of organic compounds in organic synthesis.

    Solubility
    Miscible with water.

    Notes
    Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit