Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Phenylethylamine, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
3886-69-9
C8H11N
121.183
MFCD00064405
RQEUFEKYXDPUSK-SSDOTTSWSA-N
r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine
643189
CHEBI:35322
(1R)-1-phenylethanamine
CC(C1=CC=CC=C1)N
3886-69-9
C8H11N
121.183
MFCD00064405
RQEUFEKYXDPUSK-SSDOTTSWSA-N
r-1-phenylethanamine,r-+-1-phenylethylamine,1r-1-phenylethanamine,r-1-phenylethylamine,d-alpha-methylbenzylamine,1r-1-phenylethan-1-amine,r-+-alpha-methylbenzylamine,r-alpha-methylbenzenemethanamine,unii-v022zk8gz5,r-alfa-methylbenzylamine
643189
CHEBI:35322
(1R)-1-phenylethanamine
CC(C1=CC=CC=C1)N
Chiral derivatization reagent
Specific Rotation (20°C, 589nm)%gt; +37° (neat), (20°C, 589nm) typically%gt; +39°
Packaging Glass bottle
Density 0.95g/mL
Refractive Index 1.5250 to 1.5270 (20°C, 589nm)
Enantiomeric Excess > 98.5%
Color Colorless
Quantity 500g
Infrared Spectrum Authentic
Linear Formula C6H5CH(CH3)NH2
Beilstein 12, 1092
Fieser 01,838; 02,271; 03,199; 05,441; 06,457; 07,393; 09,363; 11,411; 12,319; 13,185; 14,257
Merck Index 14, 6026
Formula Weight 121.18
Physical Form Liquid
Percent Purity ≥98.0% Total base (≥99.0% GC)
Chemical Name or Material D(+)-α-Methylbenzylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger