Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-[(S)-2-Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine, 95+%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
155830-69-6
C32H40FeP2
MFCD01074442
Josiphos SLJ002-1
155830-69-6
C32H40FeP2
MFCD01074442
Josiphos SLJ002-1
Specific Rotation Condition − 420.00 (20.00°C c=0.4,CHCl3)
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
Color Orange
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95+%
Formula Weight 542.47
Physical Form Powder
Percent Purity ≥95.0%
Chemical Name or Material (R)-(-)-1-[(S)-2-Diphenylphosphino)ferrocenyl]ethyldi-tert-butylphosphine
Show More Show Less