Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Acros Organics

Click to view available options
Quantity:
2.5g
Packaging:
Glass bottle
155806-35-2
C36H44FeP2·C2H6O
640.61
MFCD00800284
HGTBZFMPHBAUCQ-KHZPMNTOSA-N
2r-1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino ferrocene,r,s-josiphos,r-s-ppf-p cy 2,r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine,r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine ethanol adduct,r-1-sp-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine,ferrocene, 1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r,ferrocene,1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r
10984818
cyclopentane;dicyclohexyl-[(1R)-1-(2-diphenylphosphanylcyclopentyl)ethyl]phosphane;iron
CC([C]1[CH][CH][CH][C]1P(C2=CC=CC=C2)C3=CC=CC=C3)P(C4CCCCC4)C5CCCCC5.[CH]1[CH][CH][CH][CH]1.[Fe]
155806-35-2
C36H44FeP2·C2H6O
640.61
MFCD00800284
HGTBZFMPHBAUCQ-KHZPMNTOSA-N
2r-1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino ferrocene,r,s-josiphos,r-s-ppf-p cy 2,r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine,r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine ethanol adduct,r-1-sp-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine,ferrocene, 1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r,ferrocene,1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r
10984818
cyclopentane;dicyclohexyl-[(1R)-1-(2-diphenylphosphanylcyclopentyl)ethyl]phosphane;iron
CC([C]1[CH][CH][CH][C]1P(C2=CC=CC=C2)C3=CC=CC=C3)P(C4CCCCC4)C5CCCCC5.[CH]1[CH][CH][CH][CH]1.[Fe]
Specific Rotation − 350.00
Infrared Spectrum Authentic
Specific Rotation Condition - 350.00 (20.00°C c=0.4,CHCl3)
Color White
Quantity 2.5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97% min. (HPLC)
Formula Weight 640.61
Physical Form Crystals or Crystalline Powder
Percent Purity 97+%
Chemical Name or Material (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 98%
Show More Show Less