Tocris™ R 1485 dihydrochloride

Selective and high affinity 5-HT6 antagonist

Manufacturer: Tocris™ 4964/10

 View more versions of this product

Catalog No. 49-641-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors R 1485 dihydrochloride
    Molecular Weight 450.35
    Product Type R 1485 dihydrochloride
    Purity >98
    Quantity 10mg

    Selective and high affinity 5-HT6 antagonist (pKi = 8.9); displays low hERG inhibition. Exhibits >100 fold selectivity against a panel of 50 targets including other 5-HT receptor subtypes. Brain penetrant.

    • Chemical Name: 4-[(2-Fluorophenyl)sulfonyl]-3,4-dihydro-8-(1-piperazinyl)-2H-1,4-benzoxazine dihydrochloride
    • Formula: C18H20FN3O3S.2HCl
    • Solubility: Soluble to 100mM in water and to 100mM in DMSO
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit