Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-(1-Hydroxyethyl)pyridine, 98%, Acros Organics

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass Bottle
27911-63-3
C7H9NO
123.15
MFCD04972322
PPHIIIRFJKDTLG-ZCFIWIBFSA-N
r-1-pyridin-2-yl ethanol,r-2-1-hydroxyethyl pyridine,r-1-2-pyridyl ethanol,1r-1-pyridin-2-yl ethan-1-ol,r-alpha-methyl-2-pyridinemethanol,1r-1-pyridin-2-ylethanol,1r-1-pyridin-2-yl ethanol,pubchem5699,r-1-pyridin-2-yl-ethanol,1r-1-2-pyridyl ethanol
642847
(1R)-1-pyridin-2-ylethanol
CC(C1=CC=CC=N1)O
27911-63-3
C7H9NO
123.15
MFCD04972322
PPHIIIRFJKDTLG-ZCFIWIBFSA-N
r-1-pyridin-2-yl ethanol,r-2-1-hydroxyethyl pyridine,r-1-2-pyridyl ethanol,1r-1-pyridin-2-yl ethan-1-ol,r-alpha-methyl-2-pyridinemethanol,1r-1-pyridin-2-ylethanol,1r-1-pyridin-2-yl ethanol,pubchem5699,r-1-pyridin-2-yl-ethanol,1r-1-2-pyridyl ethanol
642847
(1R)-1-pyridin-2-ylethanol
CC(C1=CC=CC=N1)O
Packaging Glass Bottle
Refractive Index 1.5270 to 1.529
Specific Rotation + 24
Specific Rotation Condition +24° (31°C c=1 toluene)
Color Pink
Quantity 250mg
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 123.15
Physical Form Crystals
Percent Purity 98%
Chemical Name or Material (R)-2-(1-Hydroxyethyl)pyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning