Alfa Aesar™ (R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine

Asymmetric hydrogenation

Manufacturer: Alfa Aesar™ 4461377

Quantity 0.1g

 View more versions of this product

Catalog No. AA4461377

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 442905-33-1
    Molecular Formula C46H50N2O4P2
    Formula Weight 756.86
    MDL Number MFCD04974235
    Synonym (R)-Xylyl-P-Phos
    Chemical Name or Material (R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (di (3,5-xylyl)phosphino)-3,3'-bipyridine

    Specifications

    Formula Weight 756.86
    Molecular Formula C46H50N2O4P2
    Physical Form Powder
    CAS 442905-33-1
    Synonym (R)-Xylyl-P-Phos
    MDL Number MFCD04974235
    Melting Point 190° to 194°C
    Quantity 0.1g
    Storage Note 1
    • Packaged under Argon
    Chemical Name or Material (R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (di (3,5-xylyl)phosphino)-3,3'-bipyridine

    • Packaged under Argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit