Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine

Asymmetric hydrogenation

Manufacturer:  Alfa Aesar™ 44610-02

Quantity 0.5g

 View more versions of this product

Catalog No. AA4461002

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

221012-82-4
644.64
(R)-P-Phos
C38H34N2O4P2
MFCD04038734
(R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (diphenylphosphino)-3,3'-bipyridine

Specifications

221012-82-4
C38H34N2O4P2
MFCD04038734
(R)-P-Phos
Odorless
Powder
644.64
261° to 265°C
0.5g
(R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (diphenylphosphino)-3,3'-bipyridine

Safety and Handling

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit