Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalenechloro(p-cymene)ruthenium, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
145926-28-9,67-56-1
C54H46Cl2P2Ru+
928.88
MFCD00134456
MMAGOHNRINMYDK-UHFFFAOYSA-L
rucl p-cymene r-binap cl,r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride,chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride,r-binap rucl p-cymene cl,chloro r-+-2,2?-bis diphenylphosphino-1,1?-binaphthyl p-cymene ruthenium ii chloride
91972169
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(3+);dichloride
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].[Cl-].[Ru+3]
145926-28-9,67-56-1
C54H46Cl2P2Ru+
928.88
MFCD00134456
MMAGOHNRINMYDK-UHFFFAOYSA-L
rucl p-cymene r-binap cl,r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride,chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride,r-binap rucl p-cymene cl,chloro r-+-2,2?-bis diphenylphosphino-1,1?-binaphthyl p-cymene ruthenium ii chloride
91972169
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(3+);dichloride
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].[Cl-].[Ru+3]
Quantity 100mg
Color Brown
Infrared Spectrum Authentic
Packaging Glass bottle
NMR 96.0% min.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 928.88
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2, 2'-Bis(diphenylphosphino)-1, 1'-binaphthalenechloro(p-cymene)ruthenium chloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning