Alfa Aesar™ (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 97%

Manufacturer: Alfa Aesar™ H31791MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH31791MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 173831-50-0
    Molecular Formula C24H22O4
    Formula Weight 374.44
    MDL Number MFCD03788935
    Chemical Name or Material (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl

    Specifications

    Formula Weight 374.44
    Melting Point 101° to 105°C
    Quantity 250mg
    Optical Rotation +94.5° (C=1 in THF)
    Molecular Formula C24H22O4
    MDL Number MFCD03788935
    CAS 173831-50-0
    Chemical Name or Material (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit