Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2,2'-Diamino-1,1'-binaphthalene, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
18741-85-0
C20H16N2
284.36
MFCD00145204
DDAPSNKEOHDLKB-UHFFFAOYSA-N
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
20571
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
NC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1N
18741-85-0
C20H16N2
284.36
MFCD00145204
DDAPSNKEOHDLKB-UHFFFAOYSA-N
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
20571
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
NC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1N
Specific Rotation 156
Specific Rotation Condition 156.00 (20.00°C c=1,pyridine)
Packaging Glass bottle
Color White to Brown
Infrared Spectrum Authentic
Melting Point 242.0°C to 244.0°C
Quantity 250mg
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 284.36
Physical Form Powder
Percent Purity 99%
Chemical Name or Material (R)-(+)-2, 2'-Diamino-1, 1'-binaphthalene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning