Alfa Aesar™ (R)-(-)-2,3-O-Isopropylideneglycerol, 96%

Manufacturer: Alfa Aesar™ B2303703

Quantity 1g

 View more versions of this product

Catalog No. AAB2303703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14347-78-5
    Molecular Formula C6H12O3
    Formula Weight 132.18
    MDL Number MFCD00003213
    Beilstein 80117
    Merck Index 14,5213
    Synonym (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol
    Chemical Name or Material (R)-(-)-2,3-O-Isopropylideneglycerol

    Specifications

    Formula Weight 132.18
    Molecular Formula C6H12O3
    MDL Number MFCD00003213
    Boiling Point 82° to 83°C (14mmHg)
    Merck Index 14,5213
    Quantity 1g
    Flash Point 80°C (176°F)
    Optical Rotation −11.5° (c=5 in Methanol)
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    RTECS Number JI0400000
    CAS 14347-78-5
    Beilstein 80117
    Synonym (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Chemical Name or Material (R)-(-)-2,3-O-Isopropylideneglycerol
    Assay 96%

    GHS H Statement
    H227
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit