Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
94050-90-5
C9H10O4
182.175
MFCD00274088
AQIHDXGKQHFBNW-ZCFIWIBFSA-N
179741
(2R)-2-(4-hydroxyphenoxy)propanoic acid
CC(C(=O)O)OC1=CC=C(C=C1)O
94050-90-5
C9H10O4
182.175
MFCD00274088
AQIHDXGKQHFBNW-ZCFIWIBFSA-N
179741
(2R)-2-(4-hydroxyphenoxy)propanoic acid
CC(C(=O)O)OC1=CC=C(C=C1)O
Color White-Yellow
Quantity 5g
Formula Weight 182.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid

EINECSNumber : (3)-4259

TSCA : No