Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(+)-2-Amino-1,1-diphenyl-1-propanol, 99%

Manufacturer:  Alfa Aesar™ H5108306

Quantity 5g

 View more versions of this product

Catalog No. AAH5108306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

78603-93-7
227.3
UN3259
C15H17NO
MFCD02677967
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol

Specifications

78603-93-7
227.3
UN3259
5g
99%
C15H17NO
MFCD02677967
102° to 105°C
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit