Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Amino-6-methylheptane 95%, (98% e.e.), ACROS Organics™

Click to view available options
Quantity:
500mg
70419-11-3
C8H19N
129.247
MFCD01091018
QNIVIMYXGGFTAK-MRVPVSSYSA-N
2-heptanamine,6-methyl-, 2r,r-1,5-dimethylhexylamine,r, ?-1,5-dimethylhexylamine,r-2-amino-6-methylheptane,2r-6-methylheptan-2-amine,unii-3gq9e911bi component
6989209
(2R)-6-methylheptan-2-amine
CC(C)CCCC(C)N
70419-11-3
C8H19N
129.247
MFCD01091018
QNIVIMYXGGFTAK-MRVPVSSYSA-N
2-heptanamine,6-methyl-, 2r,r-1,5-dimethylhexylamine,r, ?-1,5-dimethylhexylamine,r-2-amino-6-methylheptane,2r-6-methylheptan-2-amine,unii-3gq9e911bi component
6989209
(2R)-6-methylheptan-2-amine
CC(C)CCCC(C)N
Color Colorless
Quantity 500mg
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Formula Weight 129.24
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (R)-2-Amino-6-methylheptane, 95%, (98% e.e.)

AIR SENSITIVE

EINECSNumber : 274-599-4

TSCA : TSCA