Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
374790-95-1
C11H15NO3
209.24
MFCD01631086
374790-95-1
C11H15NO3
209.24
MFCD01631086
Quantity 500mg
Melting Point 90.5°C
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Assay Percent Range 94% min. (HPLC)
Formula Weight 209.24
Physical Form Crystalline Powder
Percent Purity 95%,98% ee
Chemical Name or Material (R)-2-Aminobut-3-en-1-ol, benzoate salt, 95%, (98% e.e.)
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

TSCA : TSCA