Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(+)-2-(Aminomethyl)pyrrolidine dihydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
119020-04-1
C5H14Cl2N2
173.081
MFCD11974899
QZRLFKCODRAOAY-ZJIMSODOSA-N
r-pyrrolidin-2-ylmethanamine dihydrochloride,2r-pyrrolidin-2-ylmethanamine dihydrochloride,r-pyrrolidin-2-ylmethylamine dihydrochloride,r-2-pyrrolidinemethanamine 2hcl,r-pyrrolidin-2-ylmethanamine 2hcl,r-2-aminomethyl pyrrolidine dihydrochloride,2r-pyrrolidinemethanamine dihydrochloride,2r-pyrrolidin-2-yl methanamine dihydrochloride,r-2-aminomethylpyrrolidine dihydrochloride,1-2r-pyrrolidin-2-yl methanamine-hydrogen chloride 1/2
13162057
[(2R)-pyrrolidin-2-yl]methanamine;dihydrochloride
C1CC(NC1)CN.Cl.Cl
119020-04-1
C5H14Cl2N2
173.081
MFCD11974899
QZRLFKCODRAOAY-ZJIMSODOSA-N
r-pyrrolidin-2-ylmethanamine dihydrochloride,2r-pyrrolidin-2-ylmethanamine dihydrochloride,r-pyrrolidin-2-ylmethylamine dihydrochloride,r-2-pyrrolidinemethanamine 2hcl,r-pyrrolidin-2-ylmethanamine 2hcl,r-2-aminomethyl pyrrolidine dihydrochloride,2r-pyrrolidinemethanamine dihydrochloride,2r-pyrrolidin-2-yl methanamine dihydrochloride,r-2-aminomethylpyrrolidine dihydrochloride,1-2r-pyrrolidin-2-yl methanamine-hydrogen chloride 1/2
13162057
[(2R)-pyrrolidin-2-yl]methanamine;dihydrochloride
C1CC(NC1)CN.Cl.Cl
Melting Point 124°C to 130°C
Sensitivity Moisture Sensitive
Color Brown to White
Quantity 250mg
Formula Weight 173.08
Physical Form Solid
Percent Purity 95%
Chemical Name or Material (R)-(+)-2-(Aminomethyl)pyrrolidine dihydrochloride

storageNote1 : -30° to -10°C