Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Aminooctane, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass Ampule with Screw-cap
Glass bottle
34566-05-7
C8H19N
129.247
MFCD01091017
HBXNJMZWGSCKPW-MRVPVSSYSA-N
r-2-aminooctane,r-2-octylamine,2r-2-octanamine,2-octanamine, 2r,r---2-aminooctane,unii-rjm666k1lk,2r-octan-2-amine,rjm666k1lk,l-2-aminooctane,--2-octylamine
2734519
(2R)-octan-2-amine
CCCCCCC(C)N
34566-05-7
C8H19N
129.247
MFCD01091017
HBXNJMZWGSCKPW-MRVPVSSYSA-N
r-2-aminooctane,r-2-octylamine,2r-2-octanamine,2-octanamine, 2r,r---2-aminooctane,unii-rjm666k1lk,2r-octan-2-amine,rjm666k1lk,l-2-aminooctane,--2-octylamine
2734519
(2R)-octan-2-amine
CCCCCCC(C)N
Color Yellow
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3(CH2)5CH(CH3)NH2
Formula Weight 129.24
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (R)-2-Aminooctane, 95%, (98% e.e.)
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA