Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 95%

Click to view available options
Quantity:
250mg
1g
5g
131570-56-4
C23H26N2O6
426.469
MFCD00236844
MVWPBNQGEGBGRF-LJQANCHMSA-N
boc-d-dap fmoc-oh,boc-n3-fmoc-d-2,3-diaminopropionic acid,r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid,n-boc-n'-fmoc-d-2,3-diaminopropionic acid,boc-d-dpr fmoc-oh,ambotzbaa1031,boc-d-dapa fmoc-oh,pubchem14750,rarechem bk pt 0245,boc-d-dapa fmoc-oh.solvent
2755948
(2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
131570-56-4
C23H26N2O6
426.469
MFCD00236844
MVWPBNQGEGBGRF-LJQANCHMSA-N
boc-d-dap fmoc-oh,boc-n3-fmoc-d-2,3-diaminopropionic acid,r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid,n-boc-n'-fmoc-d-2,3-diaminopropionic acid,boc-d-dpr fmoc-oh,ambotzbaa1031,boc-d-dapa fmoc-oh,pubchem14750,rarechem bk pt 0245,boc-d-dapa fmoc-oh.solvent
2755948
(2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
Applications
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Quantity 5g
Solubility Information Slightly soluble in water.
Formula Weight 426.46
Percent Purity 95%
Chemical Name or Material (R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid

TSCA : No

Recommended Storage : Ambient temperatures