Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-2-Boc-aminobutyric acid dicyclohexylammonium salt, 95%

Click to view available options
Quantity:
5g
25g
27494-47-9
C21H40N2O4
384.561
MFCD00235886
HOSMYZXDFVUSCV-FCXZQVPUSA-N
boc-d-abu-oh dcha,boc-d-abu-oh.dcha,dicyclohexylamine r-2-tert-butoxycarbonyl amino butanoate,boc-d-abu-oh dicyclohexylammonium salt,2r-2-tert-butoxycarbonyl amino butanoic acid; dicha,2r-2-tert-butoxy carbonyl amino butanoic acid; n-cyclohexylcyclohexanamine,boc-d-abu-ohdcha,boc-d-abu-oh dicyclohexylammonium salt,r-n-t-butyloxycarbonyl-2-aminobutyric acid dicyclohexylamine,boc-d-abu-oh dicyclohexylammonium salt chn
2755935
N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
27494-47-9
C21H40N2O4
384.561
MFCD00235886
HOSMYZXDFVUSCV-FCXZQVPUSA-N
boc-d-abu-oh dcha,boc-d-abu-oh.dcha,dicyclohexylamine r-2-tert-butoxycarbonyl amino butanoate,boc-d-abu-oh dicyclohexylammonium salt,2r-2-tert-butoxycarbonyl amino butanoic acid; dicha,2r-2-tert-butoxy carbonyl amino butanoic acid; n-cyclohexylcyclohexanamine,boc-d-abu-ohdcha,boc-d-abu-oh dicyclohexylammonium salt,r-n-t-butyloxycarbonyl-2-aminobutyric acid dicyclohexylamine,boc-d-abu-oh dicyclohexylammonium salt chn
2755935
N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
Quantity 5g
Color White
Beilstein 6032357
Formula Weight 384.56
Physical Form Powder
Percent Purity 95%
Chemical Name or Material (R)-2-Boc-aminobutyric acid dicyclohexylammonium salt

storageNote1 : 2° to 8°C