Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Butylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
850411-21-1
C9H16O4
188.223
MFCD03427672
WXUJYEIALSHRGX-SSDOTTSWSA-N
r-2-butylsuccinic acid-1-methyl ester,3r-3-methoxycarbonylheptanoic acid,r-3-methoxycarbonyl heptanoic acid,3r-3-methoxycarbonyl heptanoic acid,butanedioic acid,2-butyl-, 1-methyl ester, 2r
5702637
(3R)-3-methoxycarbonylheptanoic acid
CCCCC(CC(=O)O)C(=O)OC
850411-21-1
C9H16O4
188.223
MFCD03427672
WXUJYEIALSHRGX-SSDOTTSWSA-N
r-2-butylsuccinic acid-1-methyl ester,3r-3-methoxycarbonylheptanoic acid,r-3-methoxycarbonyl heptanoic acid,3r-3-methoxycarbonyl heptanoic acid,butanedioic acid,2-butyl-, 1-methyl ester, 2r
5702637
(3R)-3-methoxycarbonylheptanoic acid
CCCCC(CC(=O)O)C(=O)OC
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange
Enantiomeric Excess 97.5% min. (GC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 188.2
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (R)-2-Butylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less