Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Cyclohexyl succinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
220498-07-7
C11H18O4
214.261
MFCD01091028
IYCUSOGFMJGUSU-SECBINFHSA-N
r-3-cyclohexyl-4-methoxy-4-oxobutanoic acid,r-2-cyclohexyl succinic acid-1-methyl ester,3r-3-cyclohexyl-4-methoxy-4-oxobutanoic acid,r-4-methoxy-3-cylcohexyl-4-oxobutanoic acid,pubchem23237,r-2-cyclohexylsuccinic acid 1-methyl ester,r-4-methoxy-3-cylcohexyl-4-oxobutanoicacid,3r-3-cyclohexyl-4-methoxy-4-oxo-butanoic acid,butanedioic acid,2-cyclohexyl-, 1-methyl ester, 2r
2734512
(3R)-3-cyclohexyl-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC(=O)O)C1CCCCC1
220498-07-7
C11H18O4
214.261
MFCD01091028
IYCUSOGFMJGUSU-SECBINFHSA-N
r-3-cyclohexyl-4-methoxy-4-oxobutanoic acid,r-2-cyclohexyl succinic acid-1-methyl ester,3r-3-cyclohexyl-4-methoxy-4-oxobutanoic acid,r-4-methoxy-3-cylcohexyl-4-oxobutanoic acid,pubchem23237,r-2-cyclohexylsuccinic acid 1-methyl ester,r-4-methoxy-3-cylcohexyl-4-oxobutanoicacid,3r-3-cyclohexyl-4-methoxy-4-oxo-butanoic acid,butanedioic acid,2-cyclohexyl-, 1-methyl ester, 2r
2734512
(3R)-3-cyclohexyl-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC(=O)O)C1CCCCC1
Enantiomeric Excess 97.5% min. (GC)
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 214.26
Physical Form Needle-Like Crystals
Percent Purity ≥94%
Chemical Name or Material (R)-2-Cyclohexyl succinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less