Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
130165-88-7
C12H20O4
228.288
MFCD01311785
TYYNOKUMAOAVBK-SNVBAGLBSA-N
r-2-cyclohexylmethyl succinic acid-1-methyl ester,r-2-cyclohexylmethyl succinic acid 1-methyl ester,3r-3-cyclohexylmethyl-4-methoxy-4-oxobutanoic acid,r-3-cyclohexylmethyl-4-methoxy-4-oxobutanoic acid
2734511
(3R)-3-(cyclohexylmethyl)-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1CCCCC1)CC(=O)O
130165-88-7
C12H20O4
228.288
MFCD01311785
TYYNOKUMAOAVBK-SNVBAGLBSA-N
r-2-cyclohexylmethyl succinic acid-1-methyl ester,r-2-cyclohexylmethyl succinic acid 1-methyl ester,3r-3-cyclohexylmethyl-4-methoxy-4-oxobutanoic acid,r-3-cyclohexylmethyl-4-methoxy-4-oxobutanoic acid
2734511
(3R)-3-(cyclohexylmethyl)-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC1CCCCC1)CC(=O)O
Quantity 1g
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Color Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 228.28
Physical Form Viscous Liquid
Percent Purity ≥94%
Chemical Name or Material (R)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, (98% e.e.)
Show More Show Less