Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-2-(Fmoc-amino)-4-aminobutyric acid, 95%

Click to view available options
Quantity:
250mg
1g
201484-12-0
C19H20N2O4
340.379
MFCD00237018
ZZDRDGKSMGGBDI-QGZVFWFLSA-N
fmoc-d-dab-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid,2r-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,2r-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid,n-a-fmoc-d-2,4-diaminobutyric acid,2r-4-amino-2-fmoc-amino butanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-d-2,4-diaminobutyric acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoicacid
7019710
(2R)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
201484-12-0
C19H20N2O4
340.379
MFCD00237018
ZZDRDGKSMGGBDI-QGZVFWFLSA-N
fmoc-d-dab-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid,2r-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,2r-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid,n-a-fmoc-d-2,4-diaminobutyric acid,2r-4-amino-2-fmoc-amino butanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-d-2,4-diaminobutyric acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoicacid
7019710
(2R)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
Quantity 1g
Color White
Formula Weight 340.38
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material (R)-2-(Fmoc-amino)-4-aminobutyric acid

storageNote1 : -30° to -10°C