Alfa Aesar™ (R)-(-)-2-Hydroxy-4-phenylbutyric acid, 97%

Manufacturer: Alfa Aesar™ H5280406

Quantity 5g

 View more versions of this product

Catalog No. AAH5280406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 29678-81-7
    Molecular Formula C10H12O3
    Formula Weight 180.2
    MDL Number MFCD00192219
    Beilstein 3201296
    Chemical Name or Material (R)-(-)-2-Hydroxy-4-phenylbutyric acid

    Specifications

    Formula Weight 180.2
    Molecular Formula C10H12O3
    MDL Number MFCD00192219
    Melting Point 114° to 117°C
    Quantity 5g
    Optical Rotation −9.5° (c=2.8 in Ethanol)
    CAS 29678-81-7
    Beilstein 3201296
    Chemical Name or Material (R)-(-)-2-Hydroxy-4-phenylbutyric acid
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit