Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Acros Organics

Click to view available options
Quantity:
500mg
1g
5g
Packaging:
Glass bottle
220498-08-8
C8H14O4
174.196
MFCD01091030
VHOGSWBAPDPWTG-ZCFIWIBFSA-N
r-3-methoxycarbonyl-4-methylpentanoic acid,r-2-isopropyl-succinic acid-1-methyl ester,r-2-isopropylsuccinic acid-1-methyl ester,3r-3-methoxycarbonyl-4-methylpentanoic acid,2r-2-isopropylsuccinic acid 1-methyl ester,3r-3-methoxycarbonyl-4-methylpentanoic acid,r---2-isopropylsuccinic acid 1-methyl ester,butanedioic acid,2-1-methylethyl-, 1-methyl ester, 2r
5702634
(3R)-3-methoxycarbonyl-4-methylpentanoic acid
CC(C)C(CC(=O)O)C(=O)OC
220498-08-8
C8H14O4
174.196
MFCD01091030
VHOGSWBAPDPWTG-ZCFIWIBFSA-N
r-3-methoxycarbonyl-4-methylpentanoic acid,r-2-isopropyl-succinic acid-1-methyl ester,r-2-isopropylsuccinic acid-1-methyl ester,3r-3-methoxycarbonyl-4-methylpentanoic acid,2r-2-isopropylsuccinic acid 1-methyl ester,3r-3-methoxycarbonyl-4-methylpentanoic acid,r---2-isopropylsuccinic acid 1-methyl ester,butanedioic acid,2-1-methylethyl-, 1-methyl ester, 2r
5702634
(3R)-3-methoxycarbonyl-4-methylpentanoic acid
CC(C)C(CC(=O)O)C(=O)OC
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow
Enantiomeric Excess 97.5% min. (GC)
Formula Weight 174.19
Physical Form Viscous Liquid
Percent Purity ≥94%
Chemical Name or Material (R)-2-Isopropylsuccinic acid-1-methyl ester, 98% ee