Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
112022-83-0,108-88-3
C18H20BNO
277.17
VMKAFJQFKBASMU-QGZVFWFLSA-N
r-2-methyl-cbs-oxazaborolidine,r-methyl oxazaborolidine,r-+-2-methyl-cbs-oxazaborolidine,r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine,r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole,r-me-cbs catalyst,r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole,r-me cbs,r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole,corey's catalyst
9838490
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C
112022-83-0,108-88-3
C18H20BNO
277.17
VMKAFJQFKBASMU-QGZVFWFLSA-N
r-2-methyl-cbs-oxazaborolidine,r-methyl oxazaborolidine,r-+-2-methyl-cbs-oxazaborolidine,r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine,r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole,r-me-cbs catalyst,r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole,r-me cbs,r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole,corey's catalyst
9838490
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C
Quantity 25mL
Specific Gravity 0.95
Flash Point 4°C
Density 0.9500g/mL
Packaging Glass bottle
Boiling Point 111.0°C
Solubility Information Solubility in water: reacts
Formula Weight 277.17
Chemical Name or Material (R)-(+)-2-Methyl-CBS-oxazaborolidine

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Caus

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov

GHS Signal Word: Danger