Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Methylglutaric Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
1115-81-7
C6H10O4
146.142
MFCD00671562
AQYCMVICBNBXNA-SCSAIBSYSA-N
(R)-(-)-2-Methylpentanedioic Acid
6950149
(2R)-2-methylpentanedioic acid
CC(CCC(=O)O)C(=O)O
1115-81-7
C6H10O4
146.142
MFCD00671562
AQYCMVICBNBXNA-SCSAIBSYSA-N
(R)-(-)-2-Methylpentanedioic Acid
6950149
(2R)-2-methylpentanedioic acid
CC(CCC(=O)O)C(=O)O
Color White
Melting Point 83°C
Quantity 5g
Formula Weight 146.14
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (R)-(-)-2-Methylglutaric Acid