Alfa Aesar™ (R)-(-)-2-Methylpiperazine, 98+%

Manufacturer: Alfa Aesar™ B2236601

Quantity 0.25g

 View more versions of this product

Catalog No. AAB2236601

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75336-86-6
    Molecular Formula C5H12N2
    Formula Weight 100.17
    MDL Number MFCD00192317
    UN Number UN2925
    Beilstein 3647994
    Chemical Name or Material (R)-(-)-2-Methylpiperazine

    Specifications

    Formula Weight 100.17
    Quantity 0.25g
    Optical Rotation −7° (c=1 in Ethanol)
    Molecular Formula C5H12N2
    MDL Number MFCD00192317
    CAS 75336-86-6
    UN Number UN2925
    Beilstein 3647994
    Boiling Point 155° to 156°C
    Melting Point 76° to 78°C
    Flash Point 65°C (149°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Chemical Name or Material (R)-(-)-2-Methylpiperazine
    Assay ≥98%

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit