Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-(-)-2-Methylpiperazine, 98+%

Click to view available options
Quantity:
0.25g
1g
5g
75336-86-6
C5H12N2
100.165
MFCD00192317
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
r---2-methylpiperazine,r-2-methylpiperazine,2r-2-methylpiperazine,r-mp,piperazine, 2-methyl-, 2r,r-2-methyl-piperazine,2-methylpiperazine, r,unii-p4xu643r8w
7330434
(2R)-2-methylpiperazine
CC1CNCCN1
75336-86-6
C5H12N2
100.165
MFCD00192317
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
r---2-methylpiperazine,r-2-methylpiperazine,2r-2-methylpiperazine,r-mp,piperazine, 2-methyl-, 2r,r-2-methyl-piperazine,2-methylpiperazine, r,unii-p4xu643r8w
7330434
(2R)-2-methylpiperazine
CC1CNCCN1
Sensitivity Air sensitive; Hygroscopic
Flash Point 65°C (149°F)
Melting Point 76°C to 78°C
Optical Rotation −7° (c=1 in Ethanol)
Boiling Point 155°C to 156°C
Quantity 1g
UN Number UN2925
Beilstein 3647994
Formula Weight 100.17
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Show More Show Less

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

DOTInformation : Hazard Class: 4.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures