Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Phenylglycinol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
56613-80-0
C8H12NO
138.19
MFCD00008062
IJXJGQCXFSSHNL-MRVPVSSYSA-O
r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol
2724025
(2R)-2-amino-2-phenylethanol
[NH3+][C@H](CO)C1=CC=CC=C1
56613-80-0
C8H12NO
138.19
MFCD00008062
IJXJGQCXFSSHNL-MRVPVSSYSA-O
r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol
2724025
(2R)-2-amino-2-phenylethanol
[NH3+][C@H](CO)C1=CC=CC=C1
Specific Rotation -30.5
Quantity 5g
Specific Rotation Condition −30.50° (20°C c=1,1N HCl)
Melting Point 75°C to 79°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula C6H5CH(NH2)CH2OH
Fieser 17,279
Formula Weight 137.18
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Phenylglycinol
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 260-287-5