Alfa Aesar™ (R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%

Manufacturer: Alfa Aesar™ B2467814

Quantity 25g

 View more versions of this product

Catalog No. AAB2467814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4042-36-8
    Molecular Formula C5H7NO3
    Formula Weight 129.12
    MDL Number MFCD00066212
    Beilstein 82133
    Synonym D-Pyroglutamic acid; H-D-Pyr-OH
    Chemical Name or Material (R)-(+)-2-Pyrrolidinone-5-carboxylic acid

    Specifications

    Formula Weight 129.12
    Molecular Formula C5H7NO3
    MDL Number MFCD00066212
    CAS 4042-36-8
    Beilstein 82133
    Melting Point 158° to 160°C
    Synonym D-Pyroglutamic acid; H-D-Pyr-OH
    Quantity 25g
    Optical Rotation +10.5° (c=5 in Water)
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material (R)-(+)-2-Pyrrolidinone-5-carboxylic acid
    Assay ≥98%

    Description

    Applications
    (R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

    Solubility
    Soluble in water.

    Notes
    Store away from strong oxidizing /reducing agents, strong acids, strong bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit