Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, 98%, Acros Organics

Click to view available options
Quantity:
1g
106454-69-7
C14H21NO3
251.326
MFCD00216472
LDKDMDVMMCXTMO-GFCCVEGCSA-N
n-boc-d-phenylalaninol,boc-d-phenylalaninol,r-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate,n-tert-butoxycarbonyl-d-phenylalaninol,r-2-boc-amino-3-phenyl-1-propanol,r-+-2-tert-butoxycarbonylamino-3-phenyl-1-propanol,r-2-tert-butoxycarbonylamino-3-phenyl-1-propanol,tert-butyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate,pubchem5739,r-n-tert-butoxycarbonyl-beta-phenylalaninol
7019371
tert-butyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CO
106454-69-7
C14H21NO3
251.326
MFCD00216472
LDKDMDVMMCXTMO-GFCCVEGCSA-N
n-boc-d-phenylalaninol,boc-d-phenylalaninol,r-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate,n-tert-butoxycarbonyl-d-phenylalaninol,r-2-boc-amino-3-phenyl-1-propanol,r-+-2-tert-butoxycarbonylamino-3-phenyl-1-propanol,r-2-tert-butoxycarbonylamino-3-phenyl-1-propanol,tert-butyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate,pubchem5739,r-n-tert-butoxycarbonyl-beta-phenylalaninol
7019371
tert-butyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CO
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 251.32
Chemical Name or Material (R)-(+)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, 98%