Alfa Aesar™ (R)-(-)-2-(Trifluoromethyl)pyrrolidine, 95%

Manufacturer: Alfa Aesar™ H2585303

Quantity 1g

 View more versions of this product

Catalog No. AAH2585303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 119618-29-0
    Molecular Formula C5H8F3N
    Formula Weight 139.12
    MDL Number MFCD07784180
    UN Number UN2925
    Chemical Name or Material (R)-(-)-2-(Trifluoromethyl)pyrrolidine

    Specifications

    Formula Weight 139.12
    MDL Number MFCD07784180
    UN Number UN2925
    Boiling Point 110° to 112°C
    Melting Point 27° to 31°C
    Flash Point 26°C (79°F)
    Optical Rotation −11° (c=1 in Methanol)
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    CAS 119618-29-0
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    Molecular Formula C5H8F3N
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Chemical Name or Material (R)-(-)-2-(Trifluoromethyl)pyrrolidine
    Assay 95%

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit