Learn More
(R)-3,4-DCPG, Tocris
AMPA antagonist/weak NMDA antagonist
Manufacturer:  R&D Systems 1395/50
Quantity 50mg
 View more versions of this product
Catalog No. 13-955-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>96%
239.18
(R)-3, 4-DCPG
C10H9NO6
(R)-3, 4-Dicarboxyphenylglycine
Specifications
201730-10-1
C10H9NO6
(R)-3, 4-Dicarboxyphenylglycine
(R)-3, 4-DCPG
>96%
239.18
50mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature