Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-3-Acetylthiazolidine-4-carboxylic acid, 97%

Click to view available options
Quantity:
1g
5g
54323-50-1
C6H9NO3S
175.202
MFCD00038440
WXTBYSIPOKXCPM-YFKPBYRVSA-N
r-3-acetylthiazolidine-4-carboxylic acid,4-thiazolidinecarboxylic acid, 3-acetyl-, 4r,4r-3-acetyl-1,3-thiazolidine-4-carboxylic acid,n-acetyl-l-thiaproline,r-aminofol,n-acetyl-thioproline,r-atca,acetyl-l-thiazolidine-4-carboxylic acid,r-3-acetylthiazolidin-4-carboxylic acid,r-3-acetylthiazolidine-4-carboxylicacid
197107
(4R)-3-acetyl-1,3-thiazolidine-4-carboxylic acid
CC(=O)N1CSCC1C(=O)O
54323-50-1
C6H9NO3S
175.202
MFCD00038440
WXTBYSIPOKXCPM-YFKPBYRVSA-N
r-3-acetylthiazolidine-4-carboxylic acid,4-thiazolidinecarboxylic acid, 3-acetyl-, 4r,4r-3-acetyl-1,3-thiazolidine-4-carboxylic acid,n-acetyl-l-thiaproline,r-aminofol,n-acetyl-thioproline,r-atca,acetyl-l-thiazolidine-4-carboxylic acid,r-3-acetylthiazolidin-4-carboxylic acid,r-3-acetylthiazolidine-4-carboxylicacid
197107
(4R)-3-acetyl-1,3-thiazolidine-4-carboxylic acid
CC(=O)N1CSCC1C(=O)O
Density 1.46g/mL
Boiling Point 439°C (760mmHg)
Quantity 5g
Melting Point 143°C to 145°C
Beilstein 8389
Formula Weight 175.2
Physical Form Solid
Percent Purity 97%
Chemical Name or Material (R)-3-Acetylthiazolidine-4-carboxylic acid

storageNote1 : 2° to 8°C